siRNA Details
Gene Symbol: Top2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100858.1 | NP_001094328.1 | 25 | 4130 | ||
GenBank mRNA | AB262979.1 | - | 25 | 4130 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Top2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100858.1 | NP_001094328.1 | 25 | 4130 | ||
GenBank mRNA | AB262979.1 | - | 25 | 4130 |