siRNA Details
| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108393.1 | NP_001101863.1 | 13 | 2026 | ||
| XM_006252537.2 | XP_006252599.1 | 12 | 14537 | |||
| XM_006252538.2 | XP_006252600.1 | 13 | 2073 | |||
| XM_006252539.2 | XP_006252601.1 | 13 | 2091 | |||

Zmiz1
Genomic Map