siRNA Details
| Gene Symbol: RGD1309676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014140.1 | NP_001014162.1 | 2 | 229 | ||
| XM_006252791.2 | XP_006252853.1 | 2 | 1031 | |||
| GenBank mRNA | BC079275.1 | - | 2 | 229 | 7134042 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309676| Gene Symbol: RGD1309676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014140.1 | NP_001014162.1 | 2 | 229 | ||
| XM_006252791.2 | XP_006252853.1 | 2 | 1031 | |||
| GenBank mRNA | BC079275.1 | - | 2 | 229 | 7134042 | |