siRNA Details
Gene Symbol: Ankrd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108400.2 | NP_001101870.1 | 2 | 168 | ||
GenBank mRNA | BC166884.1 | - | 2 | 168 | 6887335 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108400.2 | NP_001101870.1 | 2 | 168 | ||
GenBank mRNA | BC166884.1 | - | 2 | 168 | 6887335 |