siRNA Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341438.6 | XP_341439.3 | 7 | 1857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam149a| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341438.6 | XP_341439.3 | 7 | 1857 | ||