siRNA Details
| Gene Symbol: Purg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191554.1 | NP_001178483.1 | 1 | 874 | ||
| XM_006253233.2 | XP_006253295.1 | 1 | 872 | |||
| XM_006253234.2 | XP_006253296.1 | 1 | 872 | |||
| XM_006253235.2 | XP_006253297.1 | 1 | 872 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001098233, NM_152821, CD772246, BC116406, AF479673, and others.

Purg
Genomic Map