siRNA Details
| Gene Symbol: Bag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025130.1 | NP_001020301.1 | 613 | |||
| GenBank mRNA | BC083608.1 | - | 613 | 7133487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bag4| Gene Symbol: Bag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025130.1 | NP_001020301.1 | 613 | |||
| GenBank mRNA | BC083608.1 | - | 613 | 7133487 | ||