siRNA Details
| Gene Symbol: Golga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007731.1 | NP_001007732.1 | 1, 2 | 329 | ||
| XM_006253320.2 | XP_006253382.1 | 1, 2 | 377 | |||
| GenBank mRNA | BC078959.1 | - | 1, 2 | 329 | 7110849 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golga7| Gene Symbol: Golga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007731.1 | NP_001007732.1 | 1, 2 | 329 | ||
| XM_006253320.2 | XP_006253382.1 | 1, 2 | 377 | |||
| GenBank mRNA | BC078959.1 | - | 1, 2 | 329 | 7110849 | |