siRNA Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127301.1 | NP_001120773.1 | 19 | 2121 | ||
| XM_008771401.1 | XP_008769623.1 | 18 | 3009 | |||
| GenBank mRNA | BC166799.1 | - | 19 | 2121 | 7938235 | |
| FQ210009.1 | - | 19 | 2120 | |||

Cul4a
Genomic Map