siRNA Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253453.2 | XP_006253515.1 | 4 | 1010 | ||
| XM_006253454.2 | XP_006253516.1 | 4 | 1351 | |||
| XM_006253458.2 | XP_006253520.1 | 4 | 1450 | |||
| XM_006253459.2 | XP_006253521.1 | 2 | 1150 | |||
| XM_008771402.1 | XP_008769624.1 | 4 | 5158 | |||

Ankrd10
Genomic Map