siRNA Details
| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177684.1 | NP_001171155.1 | 3 | 349 | ||
| XM_006253460.2 | XP_006253522.1 | 3 | 797 | |||
| XM_006253461.2 | XP_006253523.1 | 3 | 795 | |||
| XM_006253462.2 | XP_006253524.1 | 3 | 788 | |||
| GenBank mRNA | BC166536.1 | - | 3 | 349 | 7191115 | |

Cars2
Genomic Map