siRNA Details
| Gene Symbol: Asb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044247.1 | NP_001037712.1 | 6 | 763 | ||
| XM_006253100.2 | XP_006253162.1 | 6 | 2238 | |||
| XM_008771246.1 | XP_008769468.1 | 6 | 1241 | |||
| XM_008771247.1 | XP_008769469.1 | 6 | 836 | |||
| XM_008771248.1 | XP_008769470.1 | 6 | 2081 | |||
| GenBank mRNA | BC098935.1 | - | 6 | 763 | 7445963 | |
| FQ214804.1 | - | 5 | 623 | |||
| FQ215658.1 | - | 6 | 680 | |||

Asb5
Genomic Map