siRNA Details
| Gene Symbol: Habp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079940.1 | NP_001073409.1 | 6, 7 | 1076 | ||
| XM_006253494.2 | XP_006253556.1 | 5, 6 | 972 | |||
| GenBank mRNA | BC129111.1 | - | 6, 7 | 1076 | 7111244 | |

Habp4
Genomic Map