siRNA Details
| Gene Symbol: PABPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135653.1 | NP_001129125.1 | 2 | 1118 | ||
| NM_001135654.1 | NP_001129126.1 | 2 | 1118 | |||
| NM_003819.3 | NP_003810.1 | 2 | 1118 | |||
| GenBank mRNA | BC029969.1 | - | 2 | 670 | ||
| BC065540.1 | - | 2 | 1021 | 6061561 | ||
| BC071591.1 | - | 2 | 1056 | 5265829 | ||
| BC094755.1 | - | 2 | 1059 | 30331896 | ||
| U33818.1 | - | 2 | 373 | |||
| U75686.1 | - | 2 | 1014 | |||
| Gene Symbol: LOC101929516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI215476.1 | - | 2 | 243 | ||

PABPC4
Genomic Map