siRNA Details
Gene Symbol: Gkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012160.1 | NP_001012160.1 | 5 | 745 | ||
XM_006253559.2 | XP_006253621.1 | 5 | 600 | |||
XM_006253561.2 | XP_006253623.1 | 4 | 522 | |||
GenBank mRNA | BC083717.1 | - | 5 | 745 | 7190699 |
