siRNA Details
| Gene Symbol: Slc25a48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771473.1 | XP_008769695.1 | 5 | 1320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a48| Gene Symbol: Slc25a48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771473.1 | XP_008769695.1 | 5 | 1320 | ||