siRNA Details
| Gene Symbol: Rab24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015023.1 | NP_001015023.1 | 4, 5 | 765 | ||
| XM_006253632.2 | XP_006253694.1 | 4, 5 | 656 | |||
| GenBank mRNA | BC091184.1 | - | 4, 5 | 765 | 7366064 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab24| Gene Symbol: Rab24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015023.1 | NP_001015023.1 | 4, 5 | 765 | ||
| XM_006253632.2 | XP_006253694.1 | 4, 5 | 656 | |||
| GenBank mRNA | BC091184.1 | - | 4, 5 | 765 | 7366064 | |