siRNA Details
| Gene Symbol: Tbc1d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108411.1 | NP_001101881.1 | 6 | 802 | ||
| XM_006253830.2 | XP_006253892.1 | 6 | 823 | |||
| XM_006253831.2 | XP_006253893.1 | 5 | 764 | |||
| XM_006253832.2 | XP_006253894.1 | 5 | 780 | |||

Tbc1d7
Genomic Map