siRNA Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014144.1 | NP_001014166.1 | 5 | 690 | ||
| XM_006253833.2 | XP_006253895.1 | 6 | 816 | |||
| GenBank mRNA | BC088287.1 | - | 5 | 690 | 7321485 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adtrp| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014144.1 | NP_001014166.1 | 5 | 690 | ||
| XM_006253833.2 | XP_006253895.1 | 6 | 816 | |||
| GenBank mRNA | BC088287.1 | - | 5 | 690 | 7321485 | |