siRNA Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037356.1 | NP_001032433.1 | 7 | 773 | ||
| GenBank mRNA | BC079236.1 | - | 7 | 753 | ||
| BC089822.1 | - | 8 | 834 | |||
| BC107922.1 | - | 7 | 773 | 7444847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pak1ip1| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037356.1 | NP_001032433.1 | 7 | 773 | ||
| GenBank mRNA | BC079236.1 | - | 7 | 753 | ||
| BC089822.1 | - | 8 | 834 | |||
| BC107922.1 | - | 7 | 773 | 7444847 | ||