siRNA Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037356.1 | NP_001032433.1 | 9, 10 | 1034 | ||
| GenBank mRNA | BC079236.1 | - | 9, 10 | 1014 | ||
| BC089822.1 | - | 10, 11 | 1095 | |||
| BC107922.1 | - | 9, 10 | 1034 | 7444847 | ||

Pak1ip1
Genomic Map