siRNA Details
| Gene Symbol: TNFRSF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297605.1 | NP_001284534.1 | 2, 3 | 484 | ||
| NM_003820.3 | NP_003811.2 | 2, 3 | 484 | |||
| XM_006711019.2 | XP_006711082.1 | 2, 3 | 260 | |||
| XM_017002720.1 | XP_016858209.1 | 2, 3 | 260 | |||
| XM_017002721.1 | XP_016858210.1 | 3, 4 | 612 | |||
| GenBank mRNA | AB208808.1 | - | 2, 3 | 278 | ||
| AF153978.1 | - | 2, 3 | 429 | |||
| AF373877.1 | - | 2, 3 | 184 | |||
| AF373878.1 | - | 2, 3 | 184 | |||
| AK124010.1 | - | 1, 2 | 29 | |||
| AK300515.1 | - | 2, 3 | 484 | |||
| AK303468.1 | - | 2, 3 | 478 | |||
| AK310076.1 | - | 2, 3 | 292 | |||
| AY358879.1 | - | 2, 3 | 265 | |||
| BC002794.2 | - | 2, 3 | 450 | 3614650 | ||
| BC018882.2 | - | 2, 3 | 450 | |||
| BX343044.2 | - | 2, 3 | 268 | |||
| CR456909.1 | - | 2, 3 | 184 | |||
| DC383433.1 | - | 2, 3 | 484 | |||
| U70321.1 | - | 2, 3 | 477 | |||
| U81232.1 | - | 2, 3 | 448 | |||

TNFRSF14
Genomic Map