siRNA Details
| Gene Symbol: Ssr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008891.2 | NP_001008891.2 | 3 | 335 | ||
| XM_006253835.2 | XP_006253897.1 | 3 | 372 | |||
| GenBank mRNA | AY321345.1 | - | 309 | |||
| BC098008.1 | - | 3 | 335 | 7454159 | ||
| FQ220080.1 | - | 3 | 303 | |||

Ssr1
Genomic Map