siRNA Details
| Gene Symbol: Sugct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014146.1 | NP_001014168.1 | 4, 5 | 321 | ||
| GenBank mRNA | BC079348.1 | - | 4, 5 | 321 | 7107655 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sugct| Gene Symbol: Sugct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014146.1 | NP_001014168.1 | 4, 5 | 321 | ||
| GenBank mRNA | BC079348.1 | - | 4, 5 | 321 | 7107655 | |