siRNA Details
| Gene Symbol: Tbce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012161.1 | NP_001012161.1 | 2 | 128 | ||
| XM_006254015.2 | XP_006254077.1 | 2 | 184 | |||
| XM_008771736.1 | XP_008769958.1 | 2 | 100 | |||
| GenBank mRNA | BC089833.1 | - | 2 | 128 | 7325371 | |

Tbce
Genomic Map