siRNA Details
| Gene Symbol: Fam208b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254220.2 | XP_006254282.1 | 14 | 4192 | ||
| XM_006254221.2 | XP_006254283.1 | 14 | 4373 | |||
| XM_006254222.2 | XP_006254284.1 | 14 | 3934 | |||
| XM_006254223.2 | XP_006254285.1 | 14 | 3862 | |||
| XM_006254224.2 | XP_006254286.1 | 14 | 3607 | |||
| XM_341552.6 | XP_341553.3 | 14 | 4193 | |||

Fam208b
Genomic Map