siRNA Details
| Gene Symbol: Dhtkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025720.1 | NP_001020891.1 | 6 | 1153 | ||
| GenBank mRNA | BC099075.1 | - | 6 | 1153 | 7441857 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhtkd1| Gene Symbol: Dhtkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025720.1 | NP_001020891.1 | 6 | 1153 | ||
| GenBank mRNA | BC099075.1 | - | 6 | 1153 | 7441857 | |