siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170343.1 | NP_001163814.1 | 12 | 1431 | ||
| XM_006254257.2 | XP_006254319.1 | 10 | 1267 | |||
| GenBank mRNA | CO567453.1 | - | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec61a2| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170343.1 | NP_001163814.1 | 12 | 1431 | ||
| XM_006254257.2 | XP_006254319.1 | 10 | 1267 | |||
| GenBank mRNA | CO567453.1 | - | 607 | |||