siRNA Details
| Gene Symbol: Cdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037543.1 | NP_001032632.1 | 4 | 569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdnf| Gene Symbol: Cdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037543.1 | NP_001032632.1 | 4 | 569 | ||