siRNA Details
| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025721.1 | NP_001020892.1 | 2, 3 | 138 | ||
| GenBank mRNA | BC097495.1 | - | 2, 3 | 138 | 7461783 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Colec12| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025721.1 | NP_001020892.1 | 2, 3 | 138 | ||
| GenBank mRNA | BC097495.1 | - | 2, 3 | 138 | 7461783 | |