siRNA Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108423.1 | NP_001101893.1 | 8 | 1862 | ||
| GenBank mRNA | FQ230531.1 | - | 5 | 638 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024182, AK004748, AK036631, AK170882, BC002255, and others.

Riok3
Genomic Map