siRNA Details
| Gene Symbol: Mocos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108425.1 | NP_001101895.1 | 13 | 2555 | ||
| XM_006254466.2 | XP_006254528.1 | 12 | 2722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mocos| Gene Symbol: Mocos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108425.1 | NP_001101895.1 | 13 | 2555 | ||
| XM_006254466.2 | XP_006254528.1 | 12 | 2722 | |||