siRNA Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012163.1 | NP_001012163.1 | 3 | 373 | ||
| XM_006254596.2 | XP_006254658.1 | 3 | 510 | |||
| XM_006254597.2 | XP_006254659.1 | 3 | 548 | |||
| XM_008772043.1 | XP_008770265.1 | 4 | 535 | |||
| GenBank mRNA | BC087108.1 | - | 3 | 373 | 7193274 | |

Lims2
Genomic Map