siRNA Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012163.1 | NP_001012163.1 | 8 | 965 | ||
| XM_006254596.2 | XP_006254658.1 | 8 | 1102 | |||
| XM_006254597.2 | XP_006254659.1 | 8 | 1140 | |||
| XM_008772043.1 | XP_008770265.1 | 9 | 1127 | |||
| GenBank mRNA | BC087108.1 | - | 8 | 965 | 7193274 | |
| FQ226177.1 | - | 4 | 316 | |||

Lims2
Genomic Map