siRNA Details
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014149.1 | NP_001014171.1 | 6 | 1137 | ||
GenBank mRNA | BC089946.1 | - | 6 | 1137 | 7311493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014149.1 | NP_001014171.1 | 6 | 1137 | ||
GenBank mRNA | BC089946.1 | - | 6 | 1137 | 7311493 |