siRNA Details
| Gene Symbol: Yipf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014150.3 | NP_001014172.3 | 6 | 804 | ||
| GenBank mRNA | BC083754.1 | - | 6 | 804 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf5| Gene Symbol: Yipf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014150.3 | NP_001014172.3 | 6 | 804 | ||
| GenBank mRNA | BC083754.1 | - | 6 | 804 | ||