siRNA Details
| Gene Symbol: Yipf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014150.3 | NP_001014172.3 | 2 | 180 | ||
| XM_006254664.2 | XP_006254726.1 | 11 | 1341 | |||
| GenBank mRNA | AY321347.1 | - | 11 | 1347 | ||
| BC083754.1 | - | 2 | 180 | |||

Yipf5
Genomic Map