siRNA Details
| Gene Symbol: Rbm27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254677.2 | XP_006254739.1 | 10 | 1701 | ||
| XM_006254678.2 | XP_006254740.1 | 9 | 1536 | |||
| XM_006254679.2 | XP_006254741.1 | 9 | 1536 | |||
| XM_006254680.2 | XP_006254742.1 | 9 | 1407 | |||
| XM_008772179.1 | XP_008770401.1 | 10 | 1701 | |||

Rbm27
Genomic Map