siRNA Details
| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008364.1 | NP_001008365.1 | 3 | 589 | ||
| XM_006254726.2 | XP_006254788.1 | 4 | 636 | |||
| XM_006254728.1 | XP_006254790.1 | 3 | 344 | |||
| XM_006254729.2 | XP_006254791.1 | 4 | 214 | |||
| XM_008772152.1 | XP_008770374.1 | 4 | 636 | |||
| XM_008772153.1 | XP_008770375.1 | 4 | 636 | |||
| GenBank mRNA | BC085883.1 | - | 3 | 589 | 7192093 | |

Snx24
Genomic Map