siRNA Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108434.1 | NP_001101904.1 | 9 | 1146 | ||
| XM_006254850.2 | XP_006254912.1 | 9 | 3400 | |||
| GenBank mRNA | FQ215467.1 | - | 1 | 30 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fech| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108434.1 | NP_001101904.1 | 9 | 1146 | ||
| XM_006254850.2 | XP_006254912.1 | 9 | 3400 | |||
| GenBank mRNA | FQ215467.1 | - | 1 | 30 | ||