siRNA Details
| Gene Symbol: LOC361346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017462.1 | NP_001017462.1 | 4 | 794 | ||
| XM_006254929.2 | XP_006254991.1 | 2 | 162 | |||
| XM_008772154.1 | XP_008770376.1 | 2 | 405 | |||
| GenBank mRNA | BC082051.1 | - | 4 | 794 | 7132090 | |

LOC361346
Genomic Map