siRNA Details
| Gene Symbol: Hdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014151.2 | NP_001014173.2 | 7 | 823 | ||
| XM_006254965.2 | XP_006255027.1 | 5 | 534 | |||
| XM_006254966.2 | XP_006255028.1 | 5 | 987 | |||
| GenBank mRNA | AY539873.1 | - | 7 | 823 | ||
| BC078941.1 | - | 5 | 508 | 7109225 | ||

Hdhd2
Genomic Map