siRNA Details
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013189.1 | NP_001013207.1 | 2 | 369 | ||
XM_006254952.2 | XP_006255014.1 | 3 | 661 | |||
XM_006254953.2 | XP_006255015.1 | 3 | 1054 | |||
XM_006254954.1 | XP_006255016.1 | 2 | 829 | |||
XM_006254955.2 | XP_006255017.1 | 3 | 660 | |||
XM_006254956.2 | XP_006255018.1 | 3 | 1051 | |||
XM_008772155.1 | XP_008770377.1 | 2 | 829 | |||
XM_008772156.1 | XP_008770378.1 | 2 | 829 | |||
GenBank mRNA | BC079072.1 | - | 2 | 827 | ||
BC088186.1 | - | 2 | 369 | 7369419 |
