siRNA Details
| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108436.1 | NP_001101906.1 | 6 | 924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo15| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108436.1 | NP_001101906.1 | 6 | 924 | ||