siRNA Details
| Gene Symbol: Ccdc102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108437.1 | NP_001101907.1 | 5 | 1259 | ||
| XM_006255116.2 | XP_006255178.1 | 5 | 1244 | |||
| XM_006255117.2 | XP_006255179.1 | 5 | 2885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc102a| Gene Symbol: Ccdc102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108437.1 | NP_001101907.1 | 5 | 1259 | ||
| XM_006255116.2 | XP_006255178.1 | 5 | 1244 | |||
| XM_006255117.2 | XP_006255179.1 | 5 | 2885 | |||