siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108438.2 | NP_001101908.1 | 8, 9 | 1152 | ||
| XM_008772314.1 | XP_008770536.1 | 6, 7 | 1244 | |||
| XM_008772315.1 | XP_008770537.1 | 7, 8 | 1336 | |||
| XM_008772316.1 | XP_008770538.1 | 7, 8 | 1333 | |||
| XM_008772317.1 | XP_008770539.1 | 7, 8 | 1092 | |||
| GenBank mRNA | BC158749.1 | - | 8, 9 | 1152 | 9101502 | |

Dok4
Genomic Map