siRNA Details
| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108440.1 | NP_001101910.1 | 2 | 137 | ||
| XM_006255267.2 | XP_006255329.1 | 3 | 605 | |||
| XM_006255268.2 | XP_006255330.1 | 3 | 605 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brd7| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108440.1 | NP_001101910.1 | 2 | 137 | ||
| XM_006255267.2 | XP_006255329.1 | 3 | 605 | |||
| XM_006255268.2 | XP_006255330.1 | 3 | 605 | |||