siRNA Details
| Gene Symbol: Gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108444.1 | NP_001101914.1 | 3 | 767 | ||
| GenBank mRNA | GQ140625.1 | - | 3 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gab1| Gene Symbol: Gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108444.1 | NP_001101914.1 | 3 | 767 | ||
| GenBank mRNA | GQ140625.1 | - | 3 | 486 | ||