siRNA Details
| Gene Symbol: Abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108446.1 | NP_001101916.1 | 7 | 687 | ||
| XM_006255407.2 | XP_006255469.1 | 7 | 704 | |||
| GenBank mRNA | AY724536.1 | - | 7 | 636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abce1| Gene Symbol: Abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108446.1 | NP_001101916.1 | 7 | 687 | ||
| XM_006255407.2 | XP_006255469.1 | 7 | 704 | |||
| GenBank mRNA | AY724536.1 | - | 7 | 636 | ||