siRNA Details
| Gene Symbol: RIOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003831.3 | NP_003822.2 | 9 | 1665 | ||
| XM_011526242.1 | XP_011524544.1 | 8 | 1537 | |||
| XM_011526243.1 | XP_011524545.1 | 9 | 1522 | |||
| GenBank mRNA | AF013591.1 | - | 9 | 1141 | ||
| AK057292.1 | - | 9 | 1665 | |||
| AK303938.1 | - | 9 | 1153 | |||
| AK304571.1 | - | 4 | 360 | |||
| AK310783.1 | - | 3 | 198 | |||
| BC028360.1 | - | 9 | 1201 | 4828351 | ||
| BC039729.1 | - | 9 | 1180 | 5745005 | ||

RIOK3
Genomic Map